missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta RI/ALK-5 Fc Chimera Protein
Click to view available options
:
50ug
Description
The Recombinant Human TGF-beta RI/ALK-5 Fc Chimera Protein is derived from NS0. The Recombinant Human TGF-beta RI/ALK-5 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Concentration | >95% by SDS-PAGE w/silver stain |
| Quantity | 50μg |
| Source | Mouse myeloma cell line,NS0-derived human TGF-beta RI/ALK-5 protein, (N-terminus) Human TGF-beta RI/ALK-5 (Ala25-Glu125) Accession &Num; P36897 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction