missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein
Click to view available options
Quantity:
1 mg
100 μg
20 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
Specifications
Specifications
| Accession Number | NP_000651.3 |
| Gene ID (Entrez) | 7040 |
| Name | TGF-beta 1 / TGFB1 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | TGFB1 |
| Biological Activity | Shows bioactivity at a concentration of 500 pg/mL in the Inhibition of IL-4-induced HT-2 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^6 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction