missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TGF-beta 1 Protein

Product Code. 18176530
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
10µg
1mg
2µg
Product Code. Quantity Formulation unitSize
18176530 2 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 2µg
18339417 2 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 2µg
18126890 10 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 10µg
18729436 1 mg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 1mg
18785756 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10µg
5 options
This item is not returnable. View return policy

Product Code. 18176530

Brand: R&D Systems™ 240B002/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.04-0.2 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 7040
Molecular Weight (g/mol) 12.8 kDa (monomer)
Name TGF-beta 1
Quantity 2 μg
Source Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Ala279-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.