missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TGF-beta 1 PLUS, Animal-Free Protein

Product Code. 18705775
Change view
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18705775 50 μg 50µg
18785845 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18705775 Supplier R&D Systems™ Supplier No. Qk0100050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human/Bovine/Porcine

No significant difference between EC50 of reference and test lots
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01137.2
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 7040
Molecular Weight (g/mol) M.W. (Observed): Dimeric TGF-beta 1 PLUS protein only, M.W. (Theroretical): 24 kDa (dimer)
Quantity 50 μg
Source E. coli-derived TGF-beta 1 protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Protein TGF-beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.