missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta 1, ACFP Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- ED50 for this effect is 0.04 to 0.2 ng/mL.
- Specific activity of recombinant human TGF-beta 1 is approximately 2.5 x 104 U/μg, which is calibrated against human TGF-beta 1 Standard (NIBSC code: 89/514).
Specifications
Specifications
| Accession Number | P01137 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 7040 |
| Molecular Weight (g/mol) | 11 kDa, reducing conditions |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda, Sf 9 (baculovirus)-derived human TGF-beta 1 protein Ala279-Ser390 Produced in an animal component free process (ACFP) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction