missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TFPI-2 Protein
Description
The Recombinant Human TFPI-2 Protein is derived from NS0. The Recombinant Human TFPI-2 Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris, NaCl and Brij-35. |
| Gene ID (Entrez) | 7980 |
| Molecular Weight (g/mol) | M.W. (Observed): 28-35 kDa, reducing conditions |
| Name | TFPI-2 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human TFPI-2 protein Asp23-Lys213,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?