missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TFAM Protein
A cDNA sequence encoding the TFAM was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13726-50ug
Additional Details : Weight : 0.01000kg
Specifications
TFAM Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSVLASCPK KPVSSYLRFS KEQLPIFKAQ NPDAKTTELI RRIAQRWREL PDSKKKIYQD AYRAEWQVYK EEISRFKEQL TPSQIMSLEK EIMDKHLKRK AMTKKKELTL LGKPKRPRSA YNVYVAERFQ EAKGDSPQEK LKTVKENWKN LSDSEKELYI QHAKEDETRY HNEMKSWEEQ MIEVGRKDLL RRTIKKQRKY GAEEC | |
Greater than 85% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TFAM protein solution (0.25 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT and 20% glycerol. |