missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TFAM Protein

A cDNA sequence encoding the TFAM was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13726-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924509

  • £99.15 / 1µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TFAM Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSSVLASCPK KPVSSYLRFS KEQLPIFKAQ NPDAKTTELI RRIAQRWREL PDSKKKIYQD AYRAEWQVYK EEISRFKEQL TPSQIMSLEK EIMDKHLKRK AMTKKKELTL LGKPKRPRSA YNVYVAERFQ EAKGDSPQEK LKTVKENWKN LSDSEKELYI QHAKEDETRY HNEMKSWEEQ MIEVGRKDLL RRTIKKQRKY GAEEC
Greater than 85% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TFAM protein solution (0.25 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers