missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TCEA1 Protein

A cDNA sequence encoding the TCEA1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13705-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934439

  • £2975.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TCEA1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMEDEVV RFAKKMDKMV QKKNAAGALD LLKELKNIPM TLELLQSTRI GMSVNAIRKQ STDEEVTSLA KSLIKSWKKL LDGPSTEKDL DEKKKEPAIT SQNSPEAREE STSSGNVSNR KDETNARDTY VSSFPRAPST SDSVRLKCRE MLAAALRTGD DYIAIGADEE ELGSQIEEAI YQEIRNTDMK YKNRVRSRIS NLKDAKNPNL RKNVLCGNIP PDLFARMTAE EMASDELKEM RKNLTKEAIR EHQMAKTGGT QTDLFTCGKC KKKNCTYTQV QTRSADEPMT TFVVCNECGN RWKFC
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TCEA1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 7.5), 1mM DTT, 50mM NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers