missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TBCB Protein

A cDNA sequence encoding the TBCB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13696-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964409

  • £2978.71 / 1mg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TBCB Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TBCB solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl and 20% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMEVTGV SAPTVTVFIS SSLNTFRSEK RYSRSLTIAE FKCKLELLVG SPASCMELEL YGVDDKFYSK LDQEDALLGS YPVDDGCRIH VIDHSGARLG EYEDVSRVEK YTISQEAYDQ RQDTVRSFLK RSKLGRYNEE ERAQQEAEAA QRLAEEKAQA SSIPVGSRCE VRAAGQSPRR GTVMYVGLTD FKPGYWIGVR YDEPLGKNDG SVNGKRYFEC QAKYGAFVKP AVVTVGDFPE EDYGLDEI
Videos
SDS
Documents
Certificates
Special Offers

Special Offers