missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TBCA Protein

A cDNA sequence encoding the TBCA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13695-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944409

  • £155.00 / 25µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TBCA Protein
Research Use Only
Recombinant Protein
E. coli
MADPRVRQIK IKTGVVKRLV KEKVMYEKEA KQQEEKIEKM RAEDGENYDI KKQAEILQES RMMIPDCQRR LEAAYLDLQR ILENEKDLEE AEEYKEARLV LDSVKLEA
Greater than 95.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The TBCA solution contains 20mM Tris-HCl buffer pH 7.5, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers