missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
An un-tagged partial Human Recombinant Tau Protein Pre-Formed Fibrils, Accession #: P10636
The Recombinant Human Tau Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page.
The Recombinant Human Tau Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page.
Specifications
Specifications
| For Use With (Application) | Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE |
| Gene ID (Entrez) | 4137 |
| Molecular Weight (g/mol) | 15.1 kDa |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Symbol | MAPT |
| Specific Reactivity | Tau |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?