missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Recombinant Human Tau 441 Active, P301S Mutant, Pre-formed Fibrils Protein [ATTO 488]

Product Code. 18389922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
Unit Size:
1 set
100µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389922 5 x 100 μg Vials 1 set
18370824 100 μg 100µg
18302502 2 x 100 μg Vials 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18389922 Supplier Novus Biologicals Supplier No. NBP314785500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Thioflavin T emission curve shows increased fluorescence (correlated to protein fibrillation) when active Tau 441 pre-formed fibrils ATTO-594 are combined with active Tau 441 monomers.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS pH 7.4
Gene ID (Entrez) 4137
Quantity 5 x 100 μg Vials
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias MAPT, microtubule-associated protein tau, MTBT1, MTBT2
Gene Symbol MAPT
Purity or Quality Grade >95%
Protein Tau 441
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.