missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAFA2/FAM19A2 protein is fully biologically active when compared to standard. The biological activity is determined by its ability to enhance neurite outgrowth of E16E18 rat embryonic cortical neurons. rHuTAFA2, immobilized at 624 μg/mL on a 96 well plate, is able to significantly enhance neurite outgrowth.
Specifications
Specifications
| Accession Number | Q8N3H0, Q8N3H0, Q8N3H0 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4. |
| Gene ID (Entrez) | 338811 |
| Molecular Weight (g/mol) | 11.2 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of TAFA2/FAM19A2 as determined by LAL method. |
| Gene Alias | Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?