missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TAF6L GST (N-Term) Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantity:
10 μg
25 μg
Conditionnement:
10µg
25µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spécification
Spécification
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 10629 |
| Molecular Weight (g/mol) | 76.9 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | FLJ11136, p300/CBP-associated factor (PCAF)-associated factor 65, PAF65-alpha, PAF65AMGC4288, PCAF-associated factor 65 alpha, PCAF-associated factor 65-alpha, TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 6L, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa |
| Gene Symbol | TAF6L |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | TAF6L |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu