missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TACI/TNFRSF13B/CVID Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
Description
Fully biologically active when compared to standard. The biological activity is determined by its ability to block human BAFF induced T2B cell survival using a concentration range of 1.0-3.0 μg/mL.
Specifications
Specifications
| For Use With (Application) | Western Blot, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 23495 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 18 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of TACI/TNFRSF13B/CVID as determined by LAL method. |
| Gene Alias | CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13 B, tumor necrosis factor receptor superfamily member 13 B, tumor necrosis factor receptor superfamily, member 13 B |
| Gene Symbol | TNFRSF13B |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction