missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human t-Plasminogen Activator/tPA His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 311-562 of Human t-Plasminogen Activator/tPA The Recombinant Human t-Plasminogen Activator/tPA Protein is derived from Baculovirus. The Recombinant Human t-Plasminogen Activator/tPA Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 50mM MES (pH 5.0) containing 5mM CaCl2, 1mM DTT, 0.5M NaCl, 20% glycerol |
| Gene ID (Entrez) | 5327 |
| Molecular Weight (g/mol) | 29.2 kDa |
| Name | Recombinant Human t-Plasminogen Activator/tPA Protein |
| Purification Method | >90%, by SDS-PAGE |
| Quantity | 50 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction