missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syntaxin-6 Protein
A DNA sequence encoding the Homo sapiens (Human) Syntaxin-6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6744-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273139.1 | |
Syntaxin-6 Protein | |
Research Use Only | |
STX6 | |
Recombinant Protein | |
E. coli | |
DPFFVVKGEV QKAVNTAQGL FQRWTELLQD PSTATREEID WTTNELRNNL RSIEWDLEDL DETISIVEAN PRKFNLDATE LSIRKAFITS TRQVVRDMKD QMSTSSVQAL AERKNRQALL GDSGSQNWST GTTDKYGRLD RELQRANSHF IEEQQAQQQL IVEQQDEQLE LVSGSIGVLK NMSQRIGGEL EEQAVMLEDF SHELESTQSR LDNVMKKLAK VSHMTSDRRQ | |
Greater than 90% as determined by SDS-PAGE. |
10228 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |