missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
An un-tagged recombinant protein corresponding to the amino acids 1-265 of Human Syntaxin 1A The Recombinant Human Syntaxin 1A Protein is derived from E. coli. The Recombinant Human Syntaxin 1A Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol, 1mM DTT |
| Gene ID (Entrez) | 6804 |
| Molecular Weight (g/mol) | TMW: 30.7kDa |
| Quantity | 0.02 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A |
| Common Name | Syntaxin 1A |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?