missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Synapsin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Synapsin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6754-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_008881.2 | |
Synapsin-1 Protein | |
Research Use Only | |
SYN1 | |
Recombinant Protein | |
E. coli | |
SRVLLVIDEP HTDWAKYFKG KKIHGEIDIK VEQAEFSDLN LVAHANGGFS VDMEVLRNGV KVVRSLKPDF VLIRQHAFSM ARNGDYRSLV IGLQYAGIPS VNSLHSVYNF CDKPWVFAQM VRLHKKLGTE EFPLIDQTFY PNHKEMLSST TYPVVVKMGH AHSGMGKVKV DNQHDFQDIA SVVALTKTYA TAEPFIDAKY DVRVQKIGQN YKAYMRTSVS GNWKTNTGSA MLEQIAMSDR YKLWVDTCSE IFGGLDICAV EALHGKDGRD HIIEVVGSSM PLIGDHQDED KQLIVELVVN KMAQALPR | |
Greater than 90.5% as determined by SDS-PAGE. |
6853 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |