missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human SULT1B1 Protein
Description
The Recombinant Human SULT1B1 Protein is derived from E. coli. The Recombinant Human SULT1B1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 27284 |
| Molecular Weight (g/mol) | 36 kDa |
| Name | Cytosolic Sulfotransferase 1B1/SULT1B1 |
| Quantity | 20 μg |
| Source | E. coli-derived human Cytosolic Sulfotransferase 1B1/SULT1B1 protein Leu2-Ile296,with an N-terminal 6-His tag Accession &Num; O43704 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?