missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SULT1A1 / STP1 Protein

A DNA sequence encoding the Homo sapiens (Human) SULT1A1 / STP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6746-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095530

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001046.2
SULT1A1 / STP1 Protein
Research Use Only
SULT1A1
Recombinant Protein
E. coli
MELIQDTSRP PLEYVKGVPL IKYFAEALGP LQSFQARPDD LLISTYPKSG TTWVSQILDM IYQGGDLEKC HRAPIFMRVP FLEFKAPGIP SGMETLKDTP APRLLKTHLP LALLPQTLLD QKVKVVYVAR NAKDVAVSYY HFYHMAKVHP EPGTWDSFLE KFMVGEVSYG SWYQHVQEWW ELSRTHPVLY LFYEDMKENP KREIQKILEF VGHSLPEETV DFVVQHTSFK EMKKNPMTNY TTVPQEFMDH SISPFMRKGM AGDWKTTFTV AQNERFDADY AEKMAGCSLS FRSEL
Greater than 80% as determined by SDS-PAGE.
6817
1 mg
Not determined.
Full Length of BC000923
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers