missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STYX Protein
A cDNA sequence encoding the STYX was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13638-HIS-5ug
Additional Details : Weight : 0.01000kg
Specifications
STYX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEDVKL EFPSLPQCKE DAEEWTYPMR REMQEILPGL FLGPYSSAMK SKLPVLQKHG ITHIICIRQN IEANFIKPNF QQLFRYLVLD IADNPVENII RFFPMTKEFI DGSLQMGGKV LVHGNAGISR SAAFVIAYIM ETFGMKYRDA FAYVQERRFCINPNAGFVHQ LQEYEAIYLA KLTIQMMSPL QIERSLSVHS GTTGSLKRTH EEEDDFGTMQ VATAQNG | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The STYX solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 1mM DTT and 0.1M NaCl. |