missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STYX Protein

A cDNA sequence encoding the STYX was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13638-HIS-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924229

  • £155.00 / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STYX Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMEDVKL EFPSLPQCKE DAEEWTYPMR REMQEILPGL FLGPYSSAMK SKLPVLQKHG ITHIICIRQN IEANFIKPNF QQLFRYLVLD IADNPVENII RFFPMTKEFI DGSLQMGGKV LVHGNAGISR SAAFVIAYIM ETFGMKYRDA FAYVQERRFCINPNAGFVHQ LQEYEAIYLA KLTIQMMSPL QIERSLSVHS GTTGSLKRTH EEEDDFGTMQ VATAQNG
Greater than 85% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The STYX solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 1mM DTT and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers