missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STX3 Protein

A cDNA sequence encoding the STX3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13635-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944219

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STX3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMMKDRL EQLKAKQLTQ DDDTDAVEIA IDNTAFMDEF FSEIEETRLN IDKISEHVEE AKKLYSIILS APIPEPKTKD DLEQLTTEIK KRANNVRNKL KSMEKHIEED EVRSSADLRI RKSQHSVLSR KFVEVMTKYN EAQVDFRERS KGRIQRQLEI TGKKTTDEEL EEMLESGNPA IFTSGIIDSQ ISKQALSEIE GRHKDIVRLE SSIKELHDMF MDIAMLVENQ GEMLDNIELN VMHTVDHVEK ARDETKKAVK YQSQARKK
Greater than 95% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
STX3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 100mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers