missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STMN2 Protein

A cDNA sequence encoding the STMN2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13621-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954169

  • £155.00 / 10µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STMN2 Protein
Research Use Only
Recombinant Protein
E. coli
MDMEVKQINK RASGQAFELI LKPPSPISEA PRTLASPKKK DLSLEEIQKK LEAAEERRKS QEAQVLKQLA EKREHEREVL QKALEENNNF SKMAEEKLIL KMEQIKENRE ANLAAIIERL QEKERHAAEV RRNKELQVEL SG
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The STMN2 protein solution contains 50mM Tris-HCl, pH 7.5 and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers