missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STK16 / PKL12 / MPSK Protein
A DNA sequence encoding the Homo sapiens (Human) STK16 / PKL12 / MPSK, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6738-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008910.1 | |
STK16 / PKL12 / MPSK Protein | |
Research Use Only | |
STK16 | |
Recombinant Protein | |
E. coli | |
GHALCVCSRG TVIIDNKRYL FIQKLGEGGF SYVDLVEGLH DGHFYALKRI LCHEQQDREE AQREADMHRL FNHPNILRLV AYCLRERGAK HEAWLLLPFF KRGTLWNEIE RLKDKGNFLT EDQILWLLLG ICRGLEAIHA KGYAHRDLKP TNILLGDEGQ PVLMDLGSMN QACIHVEGSR QALTLQDWAA QRCTISYRAP ELFSVQSHCV IDERTDVWSL GCVLYAMMFG EGPYDMVFQK GDSVALAVQN QLSIPQSPRH SSALRQLLNS MMTVDPHQRP HIPLLLSQLE ALQPPAPGQH TTQI | |
Greater than 80% as determined by SDS-PAGE. |
8576 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |