missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STK16 / PKL12 / MPSK Protein

A DNA sequence encoding the Homo sapiens (Human) STK16 / PKL12 / MPSK, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6738-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015490

  • £1735.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001008910.1
STK16 / PKL12 / MPSK Protein
Research Use Only
STK16
Recombinant Protein
E. coli
GHALCVCSRG TVIIDNKRYL FIQKLGEGGF SYVDLVEGLH DGHFYALKRI LCHEQQDREE AQREADMHRL FNHPNILRLV AYCLRERGAK HEAWLLLPFF KRGTLWNEIE RLKDKGNFLT EDQILWLLLG ICRGLEAIHA KGYAHRDLKP TNILLGDEGQ PVLMDLGSMN QACIHVEGSR QALTLQDWAA QRCTISYRAP ELFSVQSHCV IDERTDVWSL GCVLYAMMFG EGPYDMVFQK GDSVALAVQN QLSIPQSPRH SSALRQLLNS MMTVDPHQRP HIPLLLSQLE ALQPPAPGQH TTQI
Greater than 80% as determined by SDS-PAGE.
8576
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers