missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein

A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5905-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988569

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000093.1
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
Research Use Only
CYP17A1
Recombinant Protein
E. coli
GAKYPKSLLS LPLVGSLPFL PRHGHMHNNF FKLQKKYGPI YSVRMGTKTT VIVGHHQLAK EVLIKKGKDF SGRPQMATLD IASNNRKGIA FADSGAHWQL HRRLAMATFA LFKDGDQKLE KIICQEISTL CDMLATHNGQ SIDISFPVFV AVTNVISLIC FNTSYKNGDP ELNVIQNYNE GIIDNLSKDS LVDLVPWLKI FPNKTLEKLK SHVKIRNDLL NKILENYKEK FRSDSITNML DTLMQAKMNS DNGNAGPDQD SELLSDNHIL TTIGDIFGAG VETTTSVVKW TLAFLLHNPQ VKKKLYEEID QNVGFSRTPT ISDRNRLLLL EATIREVLRL RPVAPMLIPH KANVDSSIGE FAVDKGTEVI INLWALHHNE KEWHQPDQFM PERFLNPAGT QLISPSVSYL PFGAGPRSCI GEILARQELF LIMAWLLQRF DLEVPDDGQL PSLEGIPKVV FLIDSFKVKI KVRQAWREAQ AEGST
Greater than 80% as determined by SDS-PAGE.
1586
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers