missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STEAP4 Protein
A cDNA sequence encoding the STEAP4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | STEAP4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964149
|
enQuireBio™
QP13614-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954149
|
enQuireBio™
QP13614-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944149
|
enQuireBio™
QP13614-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
STEAP4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STEAP4 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMEKT CIDALPLTMN SSEKQETVCI FGTGDFGRSL GLKMLQCGYS VVFGSRNPQK TTLLPSGAEV LSYSEAAKKS GIIIIAIHRE HYDFLTELTE VLNGKILVDI SNNLKINQYP ESNAEYLAHL VPGAHVVKAF NTISAWALQS GALDASRQ | |
Greater than 90.0% as determined by SDS-PAGE. |