missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STAC Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | STAC Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMIPPSSP REDGVDGLPK EAVGAEQPPS PASTSSQESK LQKLKRSLSF KTKSLRSKSA DNFFQRTNSE DMKLQAHMVA EISPSSSPLP APGSLTSTPA RAGLHPGGKA HAFQEYIFKK PTFCDVCNHM IVGTNAKHGL RCKACKMSIH HKCTDGLAPQ RCMGKLPKGF RRYYSSPLLI HEQFGCIKEV MPIACGNKVD PVYETLRFGT SLAQRTKKGS SGSGSDSPHR TSTSDLVEVP EEANGPGGGY DLRKRSNSVF TYPENGTDDF RDPAKNINHQ GSLSKDPLQM NTYVALYKFV PQENEDLEMR PGDIITLLED SNEDWWKGKI QDRIGFFPAN FVQRLQQNEK IFRCVRTFIG CKEQGQITLK ENQICVSSEE EQDGFIRVLS GKKKGLIPLD VLENI |
| Buffer | STAC protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction