missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ST8SIA4 Protein
A DNA sequence encoding the Homo sapiens (Human) ST8SIA4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7838-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005659.1 | |
ST8SIA4 Protein | |
Research Use Only | |
ST8SIA4 | |
Recombinant Protein | |
E. coli | |
KTKEIARTEE HQETQLIGDG ELSLSRSLVN SSDKIIRKAG SSIFQHNVEG WKINSSLVLE IRKNILRFLD AERDVSVVKS SFKPGDVIHY VLDRRRTLNI SHDLHSLLPE VSPMKNRRFK TCAVVGNSGI LLDSECGKEI DSHNFVIR | |
Greater than 90% as determined by SDS-PAGE. |
7903 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |