missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ST8SIA4 Protein

A DNA sequence encoding the Homo sapiens (Human) ST8SIA4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7838-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031101

  • £384.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005659.1
ST8SIA4 Protein
Research Use Only
ST8SIA4
Recombinant Protein
E. coli
KTKEIARTEE HQETQLIGDG ELSLSRSLVN SSDKIIRKAG SSIFQHNVEG WKINSSLVLE IRKNILRFLD AERDVSVVKS SFKPGDVIHY VLDRRRTLNI SHDLHSLLPE VSPMKNRRFK TCAVVGNSGI LLDSECGKEI DSHNFVIR
Greater than 90% as determined by SDS-PAGE.
7903
100 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers