missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ST6GALNAC4 Protein
Description
Measured by its ability to transfer Neu5Ac from CMP-Neu5Ac to fetuin of fetal calf serum. The specific activity is >1000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q9H4F1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 27090 |
| Molecular Weight (g/mol) | M.W. (Observed): 36-38 kDa, reducing conditions; M.W. (theoretical): 31 kDa |
| Name | ST6GALNAC4 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human ST6GALNAC4 protein Thr36-Arg302, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?