missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ST3GAL1 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to transfer Neu5Ac from CMP-Neu5Ac to bovine B1-3 galactosyl-N-acetyl galactosamine. The specific activity is >1000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q11201 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 6482 |
| Molecular Weight (g/mol) | M.W. (Observed): 35-45 kDa, reducing conditions; M.W. (theoretical): 33.4 kDa |
| Name | ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1 |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1 protein Arg57-Arg340, with an N-terminal HA (YPYDVPDYA) tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction