missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SPR His Protein

Product Code. 18505230 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18505230

Brand: Novus Biologicals™ NBC121657100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 6697
Molecular Weight (g/mol) MolecularWeight-theroretical: 30.2 kDa
Quantity 100 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/1 μg of protein as determined by LAL method
Gene Alias EC 1.1.1.153, SDR38C1, sepiapterin reductase, sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase), short chain dehydrogenase/reductase family 38 C, member 1
Gene Symbol SPR
Research Category Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Protein SPR
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.