missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human SOX2 TAT Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Specifications
Specifications
| Accession Number | P48431, P48431, P48431 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 2 x PBS, pH 7.4, with 5% Trehalose. |
| Gene ID (Entrez) | 6657 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 36.0 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of SO x 2 AS determined by LAL method. |
| Gene Alias | ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 |
| Gene Symbol | SOX2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction