missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sorting nexin-24 Protein
A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-24, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6710-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_054754.1 | |
Sorting nexin-24 Protein | |
Research Use Only | |
SNX24 | |
Recombinant Protein | |
E. coli | |
MEVYIPSFRY EESDLERGYT VFKIEVLMNG RKHFVEKRYS EFHALHKKLK KCIKTPEIPS KHVRNWVPKV LEQRRQGLET YLQAVILENE ELPKLFLDFL NVRHLPSLPK AESCGSFDET ESEESSKLSH QPVLLFLRDP YVLPAASDFP NVVIEGVLHG IFYPHLQPR | |
Greater than 90% as determined by SDS-PAGE. |
28966 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |