missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sorting nexin-20 Protein

A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-20, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6709-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035280

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001138444.1
Sorting nexin-20 Protein
Research Use Only
SNX20
Recombinant Protein
E. coli
MASPEHPGSP GCMGPITQCT ARTQQEAPAT GPDLPHPGPD GHLDTHSGLS SNSSMTTREL QQYWQNQKCR WKHVKLLFEI ASARIEERKV SKFVVYQIIV IQ
Greater than 90% as determined by SDS-PAGE.
124460
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers