missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sorting nexin-20 Protein

A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-20, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6709-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015280

  • £1735.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138444.1
Sorting nexin-20 Protein
Research Use Only
SNX20
Recombinant Protein
E. coli
MASPEHPGSP GCMGPITQCT ARTQQEAPAT GPDLPHPGPD GHLDTHSGLS SNSSMTTREL QQYWQNQKCR WKHVKLLFEI ASARIEERKV SKFVVYQIIV IQ
Greater than 90% as determined by SDS-PAGE.
124460
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers