missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Sonic Hedgehog/Shh Protein

Product Code. 18650196 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18650196

Brand: Novus Biologicals™ NBP235265100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Sonic Hedgehog/Shh protein is fully biologically active when compared to standard. The ED50 as determined by inducing alkaline phosphatase production of murine C3H/10T1/2 cells is less than 1 μg/mL, corresponding to a specific activity of > 1.0 x 10^3 IU/mg
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15465, Q15465, Q15465
For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl.
Gene ID (Entrez) 6469
Molecular Weight (g/mol) MolecularWeight-theroretical: 19.8 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of Sonic Hedgehog/Shh as determined by LAL method.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Gene Symbol SHH
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Biologically Active Proteins
Purity or Quality Grade >98 % pure by SDS-PAGE and HPLC
Protein Sonic Hedgehog/Shh
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.