Learn More
DiscoverX Recombinant Human soluble RANK ligand
Product Code. 17322599
Encompass_Preferred
Click to view available options
:
10 μg
Product Code. 17322599
Brand: DiscoverX 921282
Recombinant Human soluble RANK ligand is an agonist for RANK. Provided as 10μg of dry powder alongside reconstitution buffer.
Soluble RANK ligand (sRANKL) is a type II protein and member of the tumor necrosis factor (TNF) superfamily which serves as an agonist for RANK. Controls bone regeneration and remodeling, and affects the immune system. Key factor in oseoclast differentiation and activation, dendritic cell survival, and T cell-dependent immune response. Provided as 10μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 10 μg
Product Type Control Ligands