missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SOD2 Protein

A cDNA sequence encoding the SOD2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10866-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945408

  • £99.15 / 5µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SOD2 Protein
Research Use Only
Specific activity is > 1,200 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and Xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume.
Human
His
SOD2 Human solution containing 20mM Tris-HCl pH 8 and 20% glycerol.
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MKHSLPDLPY DYGALEPHIN AQIMQLHHSK HHAAYVNNLN VTEEKYQEAL AKGDVTAQIA LQPALKFNGG GHINHSIFWT NLSPNGGGEP KGELLEAIKR DFGSFDKFKE KLTAASVGVQ GSGWGWLGFN KERGHLQIAA CPNQDPLQGT TGLIPLLGID VWEHAYYLQY KNVRPDYLKA IWNVINWENV TERYMACKK
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers