missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOD2 Protein
A cDNA sequence encoding the SOD2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10866-25ug
Additional Details : Weight : 0.01000kg
Specifications
SOD2 Protein | |
Research Use Only | |
Specific activity is > 1,200 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and Xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume. | |
Human | |
His | |
SOD2 Human solution containing 20mM Tris-HCl pH 8 and 20% glycerol. |
25 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKHSLPDLPY DYGALEPHIN AQIMQLHHSK HHAAYVNNLN VTEEKYQEAL AKGDVTAQIA LQPALKFNGG GHINHSIFWT NLSPNGGGEP KGELLEAIKR DFGSFDKFKE KLTAASVGVQ GSGWGWLGFN KERGHLQIAA CPNQDPLQGT TGLIPLLGID VWEHAYYLQY KNVRPDYLKA IWNVINWENV TERYMACKK | |
Greater than 95% as determined by SDS-PAGE. |