missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Recombinant Human SOD1/Cu-Zn SOD Pre-formed Fibrils Protein

Product Code. 18321415 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
Unit Size:
1 set
100µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321415 100 μg 100µg
18310837 2 x 100 μg Vials 1 set
18391646 5 x 100 μg Vials 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18321415 Supplier Novus Biologicals Supplier No. NBP314783100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PB pH 7.4, 5 mM EDTA, 50 mM DTT
Gene ID (Entrez) 6647
Quantity 100 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Gene Symbol SOD1
Research Category Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors
Purity or Quality Grade >95%, by SDS-PAGE
Protein SOD1/Cu-Zn SOD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.