missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPE Protein
A cDNA sequence encoding the SNRPE was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13549-1ug
Additional Details : Weight : 0.01000kg
Specifications
SNRPE Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAYRGQGQKV QKVMVQPINL IFRYLQNRSR IQVWLYEQVN MRIEGCIIGF DEYMNLVLDD AEEIHSKTKS RKQLGRIMLK GDNITLLQSV SN | |
Greater than 95.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SNRPE solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT, 1mM EDTA and 30% glycerol. |