missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNRPC Protein

A cDNA sequence encoding the SNRPC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13544-HIS-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943919

  • £5730.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SNRPC Protein
Research Use Only
Recombinant Protein
Baculovirus
MPKFYCDYCD TYLTHDSPSV RKTHCSGRKH KENVKDYYQK WMEEQAQSLI DKTTAAFQQG KIPPTPFSAP PPAGAMIPPP PSLPGPPRPG MMPAPHMGGP PMMPMMGPPP PGMMPVGPAP GMRPPMGGHM PMMPGPPMMR PPARPMMVPT RPGMTRPDR
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SNRPC is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers