missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human SNAPC5 GST (N-Term) Protein
Tienda Bio Techne Productos
Click to view available options
Quantity:
10 μg
25 μg
Tamaño de la unidad:
10µg
25µg
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Especificaciones
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 10302 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 33.22 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | small nuclear RNA activating complex, polypeptide 5, 19kDa, Small nuclear RNA-activating complex polypeptide 5, SNAP19small nuclear RNA activating complex, polypeptide 5, 19kD, SNAPc 19 kDa subunit, SNAPc subunit 5, snRNA-activating protein complex 19 kDa subunit, snRNA-activating protein complex subunit 5 |
| Gene Symbol | SNAPC5 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | SNAPC5 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido