missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNAP25 / SUP Protein

A DNA sequence encoding the Homo sapiens (Human) SNAP25 / SUP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP1354-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953899

  • £384.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001309831.1
SNAP25 / SUP Protein
Research Use Only
SNAP25
Recombinant Protein
E. coli
MAEDADMRNE LEEMQRRADQ LADESLESTR RMLQLVEESK DAGIRTLVML DEQGEQLERI EEGMDQINKD MKEAEKNLTD LGKFCGLCVC PCNKLKSSDA YKKAWGNNQD GVVASQPARV VDEREQMAIS GGFIRRVTND ARENEMDENL EQVSGIIGNL RHMALDMGNE IDTQNRQIDR IMEKADSNKT RIDEANQRAT KMLGSG
Greater than 90% as determined by SDS-PAGE.
6616
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers