missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNAI1 Protein
A cDNA sequence encoding the SNAI1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13529-1mg
Additional Details : Weight : 0.01000kg
Specifications
SNAI1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPRSFLV RKPSDPNRKP NYSELQDSNP EFTFQQPYDQ AHLLAAIPPP EILNPTASLP MLIWDSVLAP QAQPIAWASL RLQESPRVAE LTSLSDEDSG KGSQPPSPPS PAPSSFSSTS VSSLEAEAYA AFPGLGQVPK QLAQLSEAKD LQARKAFNCK YCNKEYLSLG ALKMHIRSHT LPCVCGTCGK AFSRPWLLQG HVRTHTGEKP FSCPHCSRAF ADRSNLRAHL QTHSDVKKYQ CQACARTFSR MSLLHKHQES GCSGCPR | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNAI1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |